accessibility menu, dialog, popup

UserName

CAS RN 850567-95-2

IUPAC Name: [4-(hydrazinecarbonyl)phenyl]boronic acid
Synonyms: 4-(hydrazinecarbonyl)phenylboronic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C7H9BN2O3
InChi Key: TZOYLNOZUCPDCL-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results