accessibility menu, dialog, popup

UserName

CAS RN 85017-80-7

LiI

CAS RN 85017-80-7

IUPAC Name: lithium(1+) hydrate iodide
Synonyms: lithium(1+) hydrate iodide
Molecular Weight (g/mol): 151.86
Molecular Formula: H2ILiO
InChi Key: WAZWGFFJLSIDMX-UHFFFAOYSA-M
SMILES: [Li+].O.[I-]
Grade 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)