accessibility menu, dialog, popup

UserName

CAS RN 85-85-8

ONHNN(Z)

CAS RN 85-85-8

IUPAC Name: (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Synonyms: (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]naphthalen-2-one
Molecular Weight (g/mol): 249.27
Molecular Formula: C15H11N3O
InChi Key: RAXUMGMWXZYADR-SDXDJHTJSA-N
SMILES: O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (7)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
Grade 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (7)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (1)
  • (3)