CAS RN 848821-61-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Alpha,Alpha-Bis[3,5-di(trifluoromethyl)phenyl]-O-(trimethylsilyl)pyrrolidine-2-methanol, TRC
CAS: 848821-61-4 Molecular Formula: C24H23F12NOSi Molecular Weight (g/mol): 597.51 Synonym: (S)-2-[Bis[3,5-bis(trifluoromethyl)phenyl](trimethylsilyloxy)methyl]pyrrolidine,(2S)-2-[Bis[3,5-bis(trifluoromethyl)phenyl](trimethylsiloxy)methyl]pyrrolidine IUPAC Name: [bis[3,5-bis(trifluoromethyl)phenyl]-[(2S)-pyrrolidin-2-yl]methoxy]-trimethylsilane SMILES: C[Si](C)(C)OC([C@@H]1CCCN1)(c2cc(cc(c2)C(F)(F)F)C(F)(F)F)c3cc(cc(c3)C(F)(F)F)C(F)(F)F