accessibility menu, dialog, popup

UserName

CAS RN 848347-44-4

NOOFFFFF

CAS RN 848347-44-4

IUPAC Name: 2,3,4,5,6-pentafluorophenyl pyridine-3-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl pyridine-3-carboxylate
Molecular Weight (g/mol): 289.16
Molecular Formula: C12H4F5NO2
InChi Key: AXHLJDBUUFUDCF-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CN=CC=C2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results