CAS RN 84832-01-9
CAS RN 84832-01-9
IUPAC Name:
methyl 2,6-difluoro-3-nitrobenzoate
Synonyms:
methyl 2,6-difluoro-3-nitrobenzoate
Molecular Weight (g/mol):
217.13
Molecular Formula:
C8H5F2NO4
InChi Key:
CIHHBTMZOLRCRL-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(F)C=CC(=C1F)[N+]([O-])=O
1
–
1
of
1
results
1
Methyl 2,6-difluoro-3-nitrobenzoate, 98%, Thermo Scientific™
CAS: 84832-01-9 Molecular Formula: C8H5F2NO4 Molecular Weight (g/mol): 217.128 MDL Number: MFCD07368850 InChI Key: CIHHBTMZOLRCRL-UHFFFAOYSA-N PubChem CID: 43449035 IUPAC Name: methyl 2,6-difluoro-3-nitrobenzoate SMILES: COC(=O)C1=C(C=CC(=C1F)[N+](=O)[O-])F