CAS RN 84793-07-7
CAS RN 84793-07-7
IUPAC Name:
5-(tert-butoxy)-4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoic acid
Synonyms:
5-(tert-butoxy)-4-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-oxopentanoic acid
Molecular Weight (g/mol):
425.48
Molecular Formula:
C24H27NO6
InChi Key:
GOPWHXPXSPIIQZ-UHFFFAOYNA-N
SMILES:
CC(C)(C)OC(=O)C(CCC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
N-Fmoc-L-glutamic acid 1-tert-butyl ester, 98%
CAS: 84793-07-7 Molecular Formula: C24H27NO6 Molecular Weight (g/mol): 425.481 MDL Number: MFCD00065648 InChI Key: GOPWHXPXSPIIQZ-FQEVSTJZSA-N Synonym: fmoc-glu-otbu,fmoc-l-glutamic acid 1-tert-butyl ester,fmoc-glu obut-oh,fmoc-glu-obut,4s-5-tert-butoxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid,s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid,fmoc-glu-ot-bu,fmoc-glu-o-t-bu,pubchem10014,ksc448a0b PubChem CID: 7017912 IUPAC Name: (4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid SMILES: CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13