accessibility menu, dialog, popup

UserName

CAS RN 846023-33-4

CH3OH2NBrHCl

CAS RN 846023-33-4

IUPAC Name: 4-bromo-3-ethoxyaniline hydrochloride
Synonyms: 4-bromo-3-ethoxyaniline hydrochloride
Molecular Weight (g/mol): 252.54
Molecular Formula: C8H11BrClNO
InChi Key: CVVLNBLBIWBEPT-UHFFFAOYSA-N
SMILES: Cl.CCOC1=C(Br)C=CC(N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)