accessibility menu, dialog, popup

UserName

SOOClFFF

CAS RN 845866-80-0

IUPAC Name: 3,3,3-trifluoropropane-1-sulfonyl chloride
Synonyms: 3,3,3-trifluoropropane-1-sulfonyl chloride
Molecular Weight (g/mol): 196.57
Molecular Formula: C3H4ClF3O2S
InChi Key: OXKHPRFGCFJUSK-UHFFFAOYSA-N
SMILES: FC(F)(F)CCS(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)