accessibility menu, dialog, popup

UserName

CAS RN 84392-17-6

OHOFFF

CAS RN 84392-17-6

IUPAC Name: 4'-(trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid
Synonyms: xenalipin xenalipinum xenalipina xenalipine 联苯利平
Molecular Weight (g/mol): 266.22
Molecular Formula: C14H9F3O2
InChi Key: IQOMYCGTGFGDFN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1C1=CC=C(C=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results