accessibility menu, dialog, popup

UserName

FHOOHOF

CAS RN 84376-20-5

IUPAC Name: 3,5-difluoro-2-hydroxybenzoic acid
Synonyms: 3,5-difluoro-2-hydroxybenzoic acid
Molecular Weight (g/mol): 174.1
Molecular Formula: C7H4F2O3
InChi Key: GZPCNALAXFNOBT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(F)=CC(F)=C1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results