accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOHO

CAS RN 84358-12-3

IUPAC Name: 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)piperidine-3-carboxylic acid
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: NXILIHONWRXHFA-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC(C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results