accessibility menu, dialog, popup

UserName

CAS RN 84315-25-3

OFF

CAS RN 84315-25-3

IUPAC Name: 5,7-difluoro-2,3-dihydro-1H-inden-1-one
Synonyms: 5,7-difluoro-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 168.14
Molecular Formula: C9H6F2O
InChi Key: XWZGNWCKXLECBO-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C2C(=O)CCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results