CAS RN 84104-30-3
CAS RN 84104-30-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R)-4-(Methylsulfinyl)-1-butylamine, TRC
CAS: 84104-30-3 Chemical Name or Material: (R)-4-(Methylsulfinyl)-1-butylamine InChI Formula: InChI=1S/C5H13NOS/c1-8(7)5-3-2-4-6/h2-6H2,1H3 Molecular Formula: C5H13NOS Molecular Weight (g/mol): 135.23 Recommended Storage: -20°C SMILES: CS(=O)CCCCN Synonym: (R)-4-(Methylsulfinyl)butylamine