accessibility menu, dialog, popup

UserName

CAS RN 84-80-0

H3CCH3CH3CH3CH3CH3OO(E)

CAS RN 84-80-0

IUPAC Name: 2-methyl-3-[(2E)-3,7,11,15-tetramethylhexadec-2-en-1-yl]-1,4-dihydronaphthalene-1,4-dione
Synonyms: vitamin k1
Molecular Weight (g/mol): 450.71
Molecular Formula: C31H46O2
InChi Key: MBWXNTAXLNYFJB-LKUDQCMENA-N
SMILES: CC(C)CCCC(C)CCCC(C)CCC\C(C)=C\CC1=C(C)C(=O)C2=CC=CC=C2C1=O
Molecular Weight (g/mol) 
  • (6)
  • (6)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (5)
Boiling Point 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (6)
  • (6)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)