accessibility menu, dialog, popup

UserName

CAS RN 84-62-8

OOOO

CAS RN 84-62-8

IUPAC Name: 1,2-diphenyl benzene-1,2-dicarboxylate
Synonyms: diphenyl phthalate
Molecular Weight (g/mol): 318.33
Molecular Formula: C20H14O4
InChi Key: DWNAQMUDCDVSLT-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=CC=C1)C1=CC=CC=C1C(=O)OC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results