CAS RN 83803-75-2
CAS RN 83803-75-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1-Chlorocyclohexyl)cyclohexyl-methanone, TRC
CAS: 83803-75-2 Molecular Formula: C13H21ClO Molecular Weight (g/mol): 228.76 Synonym: Ketone 1-Chlorocyclohexyl Cyclohexyl,(1-Chlorocyclohexyl)cyclohexylmethanone IUPAC Name: (1-chlorocyclohexyl)-cyclohexylmethanone SMILES: O=C(C1CCCCC1)C2(Cl)CCCCC2