accessibility menu, dialog, popup

UserName

CAS RN 83664-33-9

ONBr

CAS RN 83664-33-9

IUPAC Name: 2-(benzyloxy)-5-bromopyridine
Synonyms: 2-(benzyloxy)-5-bromopyridine
Molecular Weight (g/mol): 264.12
Molecular Formula: C12H10BrNO
InChi Key: LXKTVNFZAFTUNZ-UHFFFAOYSA-N
SMILES: BrC1=CN=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results