accessibility menu, dialog, popup

UserName

CAS RN 834884-61-6

SNO

CAS RN 834884-61-6

IUPAC Name: 6-(thiophen-2-yl)pyridine-3-carbaldehyde
Synonyms: 6-(thiophen-2-yl)pyridine-3-carbaldehyde
Molecular Weight (g/mol): 189.23
Molecular Formula: C10H7NOS
InChi Key: TZMYWLDEGHJHDI-UHFFFAOYSA-N
SMILES: O=CC1=CN=C(C=C1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results