accessibility menu, dialog, popup

UserName

CAS RN 832114-11-1

BOOH3CCH3H3CH3COHNNCH3CH3

CAS RN 832114-11-1

IUPAC Name: N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms: N-[2-(dimethylamino)ethyl]-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol): 318.22
Molecular Formula: C17H27BN2O3
InChi Key: KTDBEJMIGCLCSU-UHFFFAOYSA-N
SMILES: CN(C)CCNC(=O)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)