accessibility menu, dialog, popup

UserName

OHOSFFFFF

CAS RN 832-32-6

IUPAC Name: 4-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms: 4-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol): 248.17
Molecular Formula: C7H5F5O2S
InChi Key: KNXWKVQRYPQFOT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results