accessibility menu, dialog, popup

UserName

CAS RN 83-72-7

OHOO

CAS RN 83-72-7

IUPAC Name: 4-hydroxy-1,2-dihydronaphthalene-1,2-dione
Synonyms: 2-hydroxy-1,4-naphthoquinone
Molecular Weight (g/mol): 174.16
Molecular Formula: C10H6O3
InChi Key: WVCHIGAIXREVNS-UHFFFAOYSA-N
SMILES: OC1=CC(=O)C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)