accessibility menu, dialog, popup

UserName

CH3(R)(R)H3C(R)(S)(S)(S)OH(R)CH3(S)(R)CH3CH3H3C

CAS RN 83-46-5

IUPAC Name: (1R,3aS,3bS,7S,9aR,9bS,11aR)-1-[(2R,5R)-5-ethyl-6-methylheptan-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Synonyms: sitosterol harzol (24R)-ethylcholest-5-en-3b-ol 22,23-dihydrostigmasterol β-phytosterol triastonal rhamnol prostasal delta5-stigmasten-3-β-ol 谷甾醇
Molecular Weight (g/mol): 414.72
Molecular Formula: C29H50O
InChi Key: KZJWDPNRJALLNS-VJSFXXLFSA-N
SMILES: CC[C@H](CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C
Molecular Weight (g/mol) 
  • (6)
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (5)
  • (3)
Grade 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (6)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)