CAS RN 83-32-9
Acenaphthene, 99%
CAS: 83-32-9 Molecular Formula: C12H10 Molecular Weight (g/mol): 154.21 MDL Number: MFCD00003807 InChI Key: CWRYPZZKDGJXCA-UHFFFAOYSA-N Synonym: acenaphthene,1,8-ethylenenaphthalene,naphthyleneethylene,peri-ethylenenaphthalene,acenaphthylene, 1,2-dihydro,ethylenenaphthalene,1,8-dihydroacenaphthalene,unii-v8ut1gac5y,ccris 5951,v8ut1gac5y PubChem CID: 6734 ChEBI: CHEBI:22154 IUPAC Name: 1,2-dihydroacenaphthylene SMILES: C1CC2=C3C1=CC=CC3=CC=C2
Acenaphthene, 97%
CAS: 83-32-9 Molecular Formula: C12H10 Molecular Weight (g/mol): 154.21 MDL Number: MFCD00003807 InChI Key: CWRYPZZKDGJXCA-UHFFFAOYSA-N Synonym: acenaphthene,1,8-ethylenenaphthalene,naphthyleneethylene,peri-ethylenenaphthalene,acenaphthylene, 1,2-dihydro,ethylenenaphthalene,1,8-dihydroacenaphthalene,unii-v8ut1gac5y,ccris 5951,v8ut1gac5y PubChem CID: 6734 ChEBI: CHEBI:22154 IUPAC Name: 1,2-dihydroacenaphthylene SMILES: C1CC2=C3C1=CC=CC3=CC=C2
Acenaphthene (1,2-Dihydro Acenaphthylene), TRC
CAS: 83-32-9 Molecular Formula: C12 H10 Molecular Weight (g/mol): 154.21 Synonym: Acenaphthene,1,2-Dihydroacenaphthylene,peri-Ethylenenaphthalene,1,8-Ethylenenaphthalene IUPAC Name: 1,2-dihydroacenaphthylene SMILES: C1Cc2cccc3cccc1c23
EPA Method 505 Stock Standard Mixture 375 100 μg/mL in Toluene, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
HJ 478-2009 PAH Mixture 200 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
PAH Mixture 932 200 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
PAH Mixture 16 0.8-8.5 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
PAH Mixture 938 20-1000 μg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
EPA Method 610/ 8100 PAH Mixture 2000 μg/mL in Benzene:Dichloromethane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
PAH Mixture 627 0.5 μg/mL in Hexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
HJ 646-2013,HJ 805-2016,HJ 950-2018 PAH Mixture 710 200 μg/mL in Acetone:n-Hexane, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
PAH Mixture 937 500 μg/mL in Acetonitrile:Acetone:Toluene (6:3:1), Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
PAH Mixture 936 5-100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
- 1
- 2