accessibility menu, dialog, popup

UserName

CAS RN 83-12-5

OO

CAS RN 83-12-5

IUPAC Name: 2-phenyl-2,3-dihydro-1H-indene-1,3-dione
Synonyms: phenindione phenindione (substance) phenylindione rectadione merck lipha santé brand of phenindione fenhydren fenilin hemolidione emandione diadilan
Molecular Weight (g/mol): 222.24
Molecular Formula: C15H10O2
InChi Key: NFBAXHOPROOJAW-UHFFFAOYSA-N
SMILES: O=C1C(C(=O)C2=CC=CC=C12)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)