accessibility menu, dialog, popup

UserName

CAS RN 82925-02-8

CH3ONNH2OCH3

CAS RN 82925-02-8

IUPAC Name: 4-[2-(6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-yl)ethyl]aniline
Synonyms: 4-[2-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)ethyl]aniline
Molecular Weight (g/mol): 312.41
Molecular Formula: C19H24N2O2
InChi Key: DGOOLMGPMIHRFY-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2CN(CCC3=CC=C(N)C=C3)CCC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results