accessibility menu, dialog, popup

UserName

CAS RN 82894-98-2

ONOSNN

CAS RN 82894-98-2

IUPAC Name: 4-(4-nitrophenyl)-1,2,3-thiadiazole
Synonyms: 4-(4-nitrophenyl)-1,2,3-thiadiazole
Molecular Weight (g/mol): 207.21
Molecular Formula: C8H5N3O2S
InChi Key: VLRKUBATYOJPSP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C1=CSN=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results