accessibility menu, dialog, popup

UserName

CH3FF

CAS RN 82832-57-3

IUPAC Name: 4-(3,4-difluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)
Synonyms: 4-(3,4-difluorophenyl)-4'-propyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 320.47
Molecular Formula: C21H30F2
InChi Key: FSWZOZXLWVWJAH-UHFFFAOYSA-N
SMILES: CCCC1CCC(CC1)C1CCC(CC1)C1=CC(F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results