accessibility menu, dialog, popup

UserName

CH3NH3CN

CAS RN 827-36-1

IUPAC Name: 2-(dimethylamino)-2-phenylacetonitrile
Synonyms: 2-(dimethylamino)-2-phenylacetonitrile
Molecular Weight (g/mol): 160.22
Molecular Formula: C10H12N2
InChi Key: PAGHXXKYFBGJEH-UHFFFAOYNA-N
SMILES: CN(C)C(C#N)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results