CAS RN 82611-85-6
CAS RN 82611-85-6
IUPAC Name:
sodium 3-[ethyl(phenyl)amino]propane-1-sulfonate
Synonyms:
sodium 3-[ethyl(phenyl)amino]propane-1-sulfonate
Molecular Weight (g/mol):
265.3
Molecular Formula:
C11H16NNaO3S
InChi Key:
FFJBIXKLISICDT-UHFFFAOYSA-M
SMILES:
[Na+].CCN(CCCS([O-])(=O)=O)C1=CC=CC=C1
1
–
1
of
1
results
1
MedChemExpress ALPS
MedChemExpress ALPS(N-Ethyl-N-sulfopropylaniline sodium salt) is a bio-chemical reagents/chromogenic reagent.