accessibility menu, dialog, popup

UserName

CAS RN 82547-30-6

H3COBrOOH

CAS RN 82547-30-6

IUPAC Name: 3-(5-bromo-2-methoxyphenyl)propanoic acid
Synonyms: 3-(5-bromo-2-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 259.1
Molecular Formula: C10H11BrO3
InChi Key: RCLIOLQUIYTUPY-UHFFFAOYSA-N
SMILES: COC1=C(CCC(O)=O)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results