accessibility menu, dialog, popup

UserName

CAS RN 82366-55-0

H3CNCH3NOOOONOO

CAS RN 82366-55-0

IUPAC Name: 4-(dimethylamino)-3,5-dinitrobenzoate
Synonyms: 4-(dimethylamino)-3,5-dinitrobenzoate
Molecular Weight (g/mol): 254.18
Molecular Formula: C9H8N3O6
InChi Key: HPESPZDCCQMDOO-UHFFFAOYSA-M
SMILES: CN(C)C1=C(C=C(C=C1[N+]([O-])=O)C([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results