accessibility menu, dialog, popup

UserName

CAS RN 823-22-3

CH3OO

CAS RN 823-22-3

IUPAC Name: 6-methyloxan-2-one
Synonyms: hexanolactone delta-caprolactone 5-methyl-delta-valerolactone 5-hexanolide delta-hexanolactone hexalactone, delta- delta-hexanolide (RS)-delta-hexalactone 5-hydroxyhexanoic acid lactone 5-hexalactone
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: RZTOWFMDBDPERY-UHFFFAOYNA-N
SMILES: CC1CCCC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results