accessibility menu, dialog, popup

UserName

CH3SNO

CAS RN 82294-70-0

IUPAC Name: 4-methyl-1,3-thiazole-5-carbaldehyde
Synonyms: 4-methyl-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol): 127.16
Molecular Formula: C5H5NOS
InChi Key: JJVIEMFQPALZOZ-UHFFFAOYSA-N
SMILES: CC1=C(SC=N1)C=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results