accessibility menu, dialog, popup

UserName

CAS RN 822-87-7

ClO

CAS RN 822-87-7

IUPAC Name: 2-chlorocyclohexan-1-one
Synonyms: cyclohexanone, 2-chloro-
Molecular Weight (g/mol): 132.59
Molecular Formula: C6H9ClO
InChi Key: CCHNWURRBFGQCD-UHFFFAOYNA-N
SMILES: ClC1CCCCC1=O
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)