CAS RN 82061-21-0
CAS RN 82061-21-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lithium tetramethylcyclopentadienide, 97+%, Thermo Scientific™
CAS: 82061-21-0 Molecular Formula: C9H13Li MDL Number: MFCD00269824 Synonym: tetramethylcyclopentadienyl lithium,hsqylejmiwejbm-uhfffaoysa-n,1,2,3,4-tetramethylcyclopentadienyl lithium,2,3,4,5-tetramethylcyclopentadienyl lithium,lithium 1,2,3,4-tetramethyl-cyclopentadienide