accessibility menu, dialog, popup

UserName

HOOOCH2

CAS RN 818-61-1

IUPAC Name: 2-hydroxyethyl prop-2-enoate
Synonyms: 2-hydroxyethyl acrylate
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: OMIGHNLMNHATMP-UHFFFAOYSA-N
SMILES: OCCOC(=O)C=C
Boiling Point 
  • (2)
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)