accessibility menu, dialog, popup

UserName

OHOHOOOOSn

CAS RN 815-85-0

IUPAC Name: λ²-tin(2+) 2,3-dihydroxybutanedioate
Synonyms: λ²-tin(2+) tartrate
Molecular Weight (g/mol): 266.78
Molecular Formula: C4H4O6Sn
InChi Key: YXTDAZMTQFUZHK-UHFFFAOYNA-L
SMILES: [Sn++].OC(C(O)C([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results