accessibility menu, dialog, popup

UserName

CAS RN 815-84-9

IUPAC Name: λ²-lead(2+) 2,3-dihydroxybutanedioate
Synonyms: λ²-lead(2+) tartrate
Molecular Weight (g/mol): 355.3
Molecular Formula: C4H4O6Pb
InChi Key: AYPZCTCULRIASE-UHFFFAOYNA-L
SMILES: [Pb++].OC(C(O)C([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results