CAS RN 815-60-1
IUPAC Name:
(2R,4S)-2,4-dibromopentan-3-one
Synonyms:
(2R,4S)-2,4-dibromopentan-3-one
Molecular Weight (g/mol):
243.93
Molecular Formula:
C5H8Br2O
InChi Key:
UOPIOAUZQKSZRO-ZXZARUISSA-N
SMILES:
C[C@@H](Br)C(=O)[C@H](C)Br
1
–
1
of
1
results
1
2,4-Dibromo-3-pentanone, mixture of stereoisomers, 97%
CAS: 815-60-1 Molecular Formula: C5H8Br2O Molecular Weight (g/mol): 243.926 MDL Number: MFCD00015719 InChI Key: UOPIOAUZQKSZRO-UHFFFAOYSA-N Synonym: 2,4-dibromo-3-pentanone,2,4-dibromo-pentan-3-one,3-pentanone,2,4-dibromo,2,4-bis bromanyl pentan-3-one,2,4-dibromo-3-pentanone, mixture of stereoisomers PubChem CID: 11791395 IUPAC Name: 2,4-dibromopentan-3-one SMILES: CC(C(=O)C(C)Br)Br