CAS RN 811-98-3
Methanol-d4, for NMR, packaged in 0.75 ml ampoules, 99.8 atom % D
Methanol-d4, for NMR, 99.8 atom % D
CAS: 811-98-3 Molecular Formula: CH4O MDL Number: MFCD00044637 InChI Key: OKKJLVBELUTLKV-MZCSYVLQSA-N Synonym: methanol-d4,perdeuteromethanol,tetradeuteromethanol,methan-d3-ol-d,2h4 methanol,methyl alcohol-d4,cd3od,methyl-d3 alcohol-d,trideuterio deuteriooxy methane,methanol-d4, 99.8 atom % d PubChem CID: 71568
Methanol-d4, for NMR, 99.6+ atom % D
CAS: 811-98-3 Molecular Formula: CH4O Molecular Weight (g/mol): 36.066 MDL Number: MFCD00044637 InChI Key: OKKJLVBELUTLKV-MZCSYVLQSA-N Synonym: methanol-d4,perdeuteromethanol,tetradeuteromethanol,methan-d3-ol-d,2h4 methanol,methyl alcohol-d4,cd3od,methyl-d3 alcohol-d,trideuterio deuteriooxy methane,methanol-d4, 99.8 atom % d PubChem CID: 71568 IUPAC Name: trideuterio(deuteriooxy)methane SMILES: CO
Methanol-d4, for NMR, 99.5 atom % D
CAS: 811-98-3 Molecular Formula: CH4O Molecular Weight (g/mol): 36.066 MDL Number: MFCD00044637 InChI Key: OKKJLVBELUTLKV-MZCSYVLQSA-N Synonym: methanol-d4,perdeuteromethanol,tetradeuteromethanol,methan-d3-ol-d,2h4 methanol,methyl alcohol-d4,cd3od,methyl-d3 alcohol-d,trideuterio deuteriooxy methane,methanol-d4, 99.8 atom % d PubChem CID: 71568 IUPAC Name: trideuterio(deuteriooxy)methane SMILES: CO
Methanol-d4, for NMR, contains 0.03 v/v% TMS, 99.8 atom % D
CAS: 811-98-3 Molecular Formula: CH4O MDL Number: MFCD00044637 InChI Key: OKKJLVBELUTLKV-MZCSYVLQSA-N Synonym: trideuterio deuteriooxy methane,perdeuteromethanol,methyl-d3 alcohol-d,methan-d3-ol-d,methyl alcohol-d4,tetradeuteromethanol,2h4 methanol,cd3od,methanol-d4,methanol-d4, 99.8 atom % d PubChem CID: 71568
Methanol-d4, for NMR, packaged in 0.75 ml ampoules, 100.0 atom % D
Methanol-d{4}, 99.8% (Isotopic)
CAS: 811-98-3 Molecular Formula: CH4O Molecular Weight (g/mol): 36.066 MDL Number: MFCD00044637 InChI Key: OKKJLVBELUTLKV-MZCSYVLQSA-N Synonym: methanol-d4,perdeuteromethanol,tetradeuteromethanol,methan-d3-ol-d,2h4 methanol,methyl alcohol-d4,cd3od,methyl-d3 alcohol-d,trideuterio deuteriooxy methane,methanol-d4, 99.8 atom % d PubChem CID: 71568 IUPAC Name: trideuterio(deuteriooxy)methane SMILES: CO
Methanol-d{4}, 99.8%(Isotopic)
CAS: 811-98-3 Molecular Formula: CH4O Molecular Weight (g/mol): 36.066 MDL Number: MFCD00044637 InChI Key: OKKJLVBELUTLKV-MZCSYVLQSA-N Synonym: methanol-d4,perdeuteromethanol,tetradeuteromethanol,methan-d3-ol-d,2h4 methanol,methyl alcohol-d4,cd3od,methyl-d3 alcohol-d,trideuterio deuteriooxy methane,methanol-d4, 99.8 atom % d PubChem CID: 71568 IUPAC Name: trideuterio(deuteriooxy)methane SMILES: CO
Methanol-d4, for NMR, with 0.03% TMS, in 0.75 ml ampoules, 99.8 atom % D
Methanol-d{4}, 100%(Isotopic)
CAS: 811-98-3 Molecular Formula: CH4O Molecular Weight (g/mol): 36.066 MDL Number: MFCD00044637 InChI Key: OKKJLVBELUTLKV-MZCSYVLQSA-N Synonym: methanol-d4,perdeuteromethanol,tetradeuteromethanol,methan-d3-ol-d,2h4 methanol,methyl alcohol-d4,cd3od,methyl-d3 alcohol-d,trideuterio deuteriooxy methane,methanol-d4, 99.8 atom % d PubChem CID: 71568 IUPAC Name: trideuterio(deuteriooxy)methane SMILES: CO
Methanol-d{4}, 99.8% (Isotopic), contains 0.05% v/v TMS
CAS: 811-98-3 Molecular Formula: CH4O Molecular Weight (g/mol): 36.066 MDL Number: MFCD00044637 InChI Key: OKKJLVBELUTLKV-MZCSYVLQSA-N Synonym: methanol-d4,perdeuteromethanol,tetradeuteromethanol,methan-d3-ol-d,2h4 methanol,methyl alcohol-d4,cd3od,methyl-d3 alcohol-d,trideuterio deuteriooxy methane,methanol-d4, 99.8 atom % d PubChem CID: 71568 IUPAC Name: trideuterio(deuteriooxy)methane SMILES: CO
Methyl Alcohol-d4, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
- 1
- 2