accessibility menu, dialog, popup

UserName

CAS RN 81045-39-8

BrBrN

CAS RN 81045-39-8

IUPAC Name: 5,8-dibromoisoquinoline
Synonyms: 5,8-dibromoisoquinoline
Molecular Weight (g/mol): 286.95
Molecular Formula: C9H5Br2N
InChi Key: KNBALCRXZUTMBP-UHFFFAOYSA-N
SMILES: BrC1=C2C=CN=CC2=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results