accessibility menu, dialog, popup

UserName

CAS RN 81-13-0

CH3H3CHO(R)OHOHNOH

CAS RN 81-13-0

IUPAC Name: (2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Synonyms: dexpanthenol pantothenol ilopan
Molecular Weight (g/mol): 205.25
Molecular Formula: C9H19NO4
InChi Key: SNPLKNRPJHDVJA-ZETCQYMHSA-N
SMILES: CC(C)(CO)[C@@H](O)C(=O)NCCCO
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)