accessibility menu, dialog, popup

UserName

CAS RN 80822-15-7

OO(S)(S)OHOOOOHOH2O

CAS RN 80822-15-7

IUPAC Name: 2,3-bis(benzoyloxy)butanedioic acid hydrate
Synonyms: 2,3-bis(benzoyloxy)butanedioic acid hydrate
Molecular Weight (g/mol): 376.32
Molecular Formula: C18H16O9
InChi Key: DXDIHODZARUBLA-UHFFFAOYNA-N
SMILES: O.OC(=O)C(OC(=O)C1=CC=CC=C1)C(OC(=O)C1=CC=CC=C1)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results