CAS RN 80082-65-1
CAS RN 80082-65-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(2S,3S)-3-Amino-2-methyl-4-oxoazetidine-1-sulphonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2S,3S)-3-Amino-2-methyl-4-oxoazetidine-1-sulphonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.