accessibility menu, dialog, popup

UserName

CAS RN 8008-63-7

H3COOHHOCH3OHOHCH3

CAS RN 8008-63-7

IUPAC Name: 4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanoic acid
Synonyms: acid, cholic ca - cholic acid cholalic acid cholalate cholalin cholic acid, 5β- 5β-cholic acid colalin 5b-cholic acid 5b-cholate
Molecular Weight (g/mol): 408.58
Molecular Formula: C24H40O5
InChi Key: BHQCQFFYRZLCQQ-UHFFFAOYNA-N
SMILES: CC(CCC(O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)