CAS RN 8001-25-0
CAS RN 8001-25-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
7
of
7
results
1
2
Olive oil, pure, virgin
CAS: 8001-25-0
4
Promotions Available
Olive Oil Reagent MP Biomedicals
Olive Oil Reagent MP Biomedicals
5
Olive Oil, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
6
Olive Oil, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
7
Olive Oil, Pure, Fisher Chemical™
CAS: 8001-25-0 MDL Number: 131764