accessibility menu, dialog, popup

UserName

CAS RN 80-69-3

IUPAC Name: 2-hydroxypropanedioic acid
Synonyms: tartronic acid
Molecular Weight (g/mol): 120.06
Molecular Formula: C3H4O5
InChi Key: ROBFUDYVXSDBQM-UHFFFAOYSA-N
SMILES: OC(C(O)=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results