accessibility menu, dialog, popup

UserName

CAS RN 80-07-9

IUPAC Name: 1-chloro-4-(4-chlorobenzenesulfonyl)benzene
Synonyms: 4,4'-dichlorodiphenyl sulfone
Molecular Weight (g/mol): 287.15
Molecular Formula: C12H8Cl2O2S
InChi Key: GPAPPPVRLPGFEQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (5)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)