accessibility menu, dialog, popup

UserName

SBrOH

CAS RN 79757-77-0

IUPAC Name: (4-bromothiophen-2-yl)methanol
Synonyms: (4-bromothiophen-2-yl)methanol
Molecular Weight (g/mol): 193.06
Molecular Formula: C5H5BrOS
InChi Key: PXZNJHHUYJRFPZ-UHFFFAOYSA-N
SMILES: OCC1=CC(Br)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results